Invitrogen
Manufacturer Code:PA585211
Catalog # PIPA585211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human ADAMTS4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM-TS 4; ADAMTS-2 antibody ADAMTS-4 antibody ADMP-1 antibody KIAA0688 antibody ADAMTS4 antibody ADAM-TS4 antibody a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 4 antibody A disintegrin and metalloproteinase with thrombospondin motifs 4 antibody aggrecanase-1 antibody ADAM-TS 4 antibody ADAM metallopeptidase with thrombospondin type 1 motif 4 antibody; ADAMTS-4; Aggrecanase-1
Gene Aliases: 3830423K05; ADAM-TS4; ADAMTS-2; Adamts4; ADMP-1; mKIAA0688
UniProt ID: (Mouse) Q8BNJ2
Entrez Gene ID: (Mouse) 240913
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.