Invitrogen
Manufacturer Code:PA11749A
Catalog # 01-670-146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C V(394) M A H V D P E E P(403) of mouse ADAMTS4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAM-TS 4; ADAM-TS4; ADAMTS family member; ADAMTS-4; ADMP-1; Aggrecanase 1 A disintegrin and metalloproteinase with thrombospondin motifs 4 ADAM-TS 4 ADAM-TS4 ADMP-1; Aggrecanase-1; metalloproteinase
Gene Aliases: 3830423K05; ADAM-TS4; ADAMTS-2; ADAMTS-4; ADAMTS4; ADMP-1; KIAA0688; mKIAA0688; UNQ769/PRO1563
UniProt ID: (Bovine) Q9TT93, (Human) O75173, (Mouse) Q8BNJ2
Entrez Gene ID: (Bovine) 286806, (Human) 9507, (Pig) 100157276, (Mouse) 240913
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.