Novus Biologicals
Manufacturer Code:NBP2843930.1ML
Catalog # NB032321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle terminal region of human ADAMTS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 2; A disintegrin and metalloproteinase with thrombospondin motifs 2 a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif 2 ADAM metallopeptidase with thrombospondin type 1 motif 2 ADAM-TS 2 ADAMTS-2 ADAM-TS2EC 3.4.24.14 ADAMTS-3 EC 3.4.24 NPIDKFZp686F12218 PC I-NP PCI-NP PCINPhPCPNI PCPNI pNPI Procollagen I N-proteinase Procollagen I/II amino propeptide-processing enzyme Procollagen N-endopeptidase; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 2; ADAM-TS 2; PC I-NP; pNPI; Procollagen I N-proteinase; Procollagen I/II amino propeptide-processing enzyme; Procollagen N-endopeptidase
Gene Aliases: ADAM-TS2; ADAMTS-2; ADAMTS-3; ADAMTS2; NPI; PC I-NP; PCI-NP; PCINP; PCPNI; PNPI
UniProt ID: (Human) O95450
Entrez Gene ID: (Human) 9509
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.