R&D Systems
Manufacturer Code:MAB4245SP
Catalog # MAB4245SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 442323 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ADAMTS13 isoform 1 Gln34-Trp688 (predicted) Accession # Q76LX8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 13; A disintegrin and metalloproteinase with thrombospondin motifs 13 a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif 13 ADAM metallopeptidase with thrombospondin type 1 motif 13 ADAM-TS13 ADAMTS-13 DKFZp434C2322 EC 3.4.24.14 EC 3.4.24.82 EC 3.4.24.87 FLJ42993 MGC118899 MGC118900 TTP TTPADAM-TS 13 vWF-cleaving protease vWF-CP vWF-CPC9orf8 VWFCPvon Willebrand factor-cleaving protease; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAM-TS 13; von Willebrand factor-cleaving protease; vWF-cleaving protease; vWF-CP
Gene Aliases: ADAM-TS13; ADAMTS-13; ADAMTS13; C9orf8; UNQ6102/PRO20085; vWF-CP; VWFCP
UniProt ID: (Human) Q76LX8
Entrez Gene ID: (Human) 11093
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.