Invitrogen
Manufacturer Code:PA578727
Catalog # PIPA578727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human ADAMTS13 recombit protein (Position: A299-R488). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 13; A disintegrin and metalloproteinase with thrombospondin motifs 13 ADAM-TS 13 ADAM-TS13 ADAMTS-13; A disintegrin and metalloproteinase with thrombospondin motifs 13-like; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAM-TS 13; von Willebrand factor-cleaving protease; vWF-cleaving protease; vWF-CP; vWF-CP mRNA for von Willebrand factor-cleaving
Gene Aliases: ADAM-TS13; ADAMTS-13; ADAMTS13; C9orf8; Gm710; UNQ6102/PRO20085; vWF-CP; VWFCP
UniProt ID: (Human) Q76LX8, (Mouse) Q769J6
Entrez Gene ID: (Human) 11093, (Mouse) 279028, (Rat) 102554393
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.