Invitrogen
Manufacturer Code:PA5111164
Catalog # PIPA5111164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ADAMTS13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase A disintegrin and metalloproteinase with thrombospondin motifs 13 ADAM ADAMs ADAM-TS 13 Adamts13 ADAM-TS13 ADAMTS-13 C9orf8 Gm710 metalloendopeptidases UNQ6102/PRO20085 von Willebrand factor-cleaving protease VWFCP vWF-CP; A disintegrin and metalloproteinase with thrombospondin motifs 13; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 13; ADAM metallopeptidase with thrombospondin type 1 motif, 13; ADAM-TS 13; von Willebrand factor-cleaving protease; vWF-cleaving protease; vWF-CP
Gene Aliases: ADAM-TS13; ADAMTS-13; ADAMTS13; C9orf8; UNQ6102/PRO20085; vWF-CP; VWFCP
UniProt ID: (Human) Q76LX8
Entrez Gene ID: (Human) 11093
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.