Invitrogen
Manufacturer Code:PA557489
Catalog # PIPA557489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADAMTS12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 12; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 12; ADAM metallopeptidase with thrombospondin type 1 motif, 12; ADAM-TS 12; ADAM-TS12; ADAMTS-12; UNQ1918 PRO4389
Gene Aliases: ADAMTS12; PRO4389; UNQ1918/PRO4389
UniProt ID: (Human) A2RRN9
Entrez Gene ID: (Human) 81792
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.