Novus Biologicals
Manufacturer Code:NB10075608
Catalog # NB10075608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody was a sequence from 297a.a. to 446a.a. from ADAMTS12(a 1593a.a. long protein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 12; A disintegrin and metalloproteinase with thrombospondin motifs 12 a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif 12 ADAM metallopeptidase with thrombospondin type 1 motif 12 ADAM-TS 12 ADAM-TS12 ADAMTS-12 EC 3.4.24 EC 3.4.24.- EC 3.4.24.82 PRO4389; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 12; ADAM metallopeptidase with thrombospondin type 1 motif, 12; ADAM-TS 12; ADAM-TS12; ADAMTS-12
Gene Aliases: ADAMTS12; PRO4389; UNQ1918/PRO4389
UniProt ID: (Human) A2RRN9
Entrez Gene ID: (Human) 81792
Molecular Function:
enzyme modulator
extracellular matrix glycoprotein
extracellular matrix protein
hydrolase
metalloprotease
protease
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.