OriGene
Manufacturer Code:CF804503
Catalog # 50-166-7017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4H2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 412-644 of human ADAMTS1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A disintegrin and metalloproteinase with thrombospondin motifs 1; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1; ADAM metallopeptidase with thrombospondin type 1 motif, 1; ADAM-TS 1; ADAM-TS1; ADAMTS-1; C3-C5 METH1; human metalloproteinase with thrombospondin type 1 motifs; METH-1
Gene Aliases: ADAMTS1; C3-C5; KIAA1346; METH1
UniProt ID: (Human) Q9UHI8
Entrez Gene ID: (Human) 9510
Molecular Function: enzyme modulator extracellular matrix glycoprotein extracellular matrix protein hydrolase metalloprotease protease protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.