Invitrogen
Manufacturer Code:PA525959
Catalog # PIPA525959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 704-733 amino acids from the C-terminal region of human ADAM9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAM 9; ADAM metallopeptidase domain 9 (meltrin gamma); Cellular disintegrin-related protein; cone rod dystrophy 9; CORD9 MCMP MDC9 Mltng ADAM metallopeptidase domain 9 (meltrin gamma) cellular disintegrin-related protein cone rod dystrophy 9 disintegrin and metalloproteinase domain-containing protein 9 metalloprotease/disintegrin/cysteine-rich protein 9 myeloma cell metalloproteinase; Disintegrin and metalloproteinase domain-containing protein 9; Meltrin-gamma; Metalloprotease/disintegrin/cysteine-rich protein 9; Myeloma cell metalloproteinase
Gene Aliases: ADAM9; CORD9; KIAA0021; MCMP; MDC9; MLTNG
UniProt ID: (Human) Q13443
Entrez Gene ID: (Human) 8754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.