Novus Biologicals
Manufacturer Code:NBP159010
Catalog # NBP159010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ADAM9(ADAM metallopeptidase domain 9 (meltrin gamma)) The peptide sequence was selected from the middle region of ADAM9. Peptide sequence NKGGNCLLNIPKPDEAYSAPSCGNKLVDAGEECDCGTPKECELDPCCEGS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 9 (meltrin gamma) ADAM 9 ADAM metallopeptidase domain 9 ADAM metallopeptidase domain 9 (meltrin gamma) Cellular disintegrin-related protein cone rod dystrophy 9 CORD9 disintegrin and metalloproteinase domain-containing protein 9 EC 3.4.24 EC 3.4.24.- MCMPMDC9KIAA0021Mltng Meltrin-gamma Metalloprotease/disintegrin/cysteine-rich protein 9 MLTNG Myeloma cell metalloproteinase; ADAM 9; ADAM metallopeptidase domain 9 (meltrin gamma); Cellular disintegrin-related protein; cone rod dystrophy 9; Disintegrin and metalloproteinase domain-containing protein 9; Meltrin-gamma; Metalloprotease/disintegrin/cysteine-rich protein 9; Myeloma cell metalloproteinase
Gene Aliases: ADAM9; CORD9; KIAA0021; MCMP; MDC9; MLTNG
UniProt ID: (Human) Q13443
Entrez Gene ID: (Human) 8754
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.