R&D Systems
Manufacturer Code:MAB10311
Catalog # MAB10311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 143338 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ADAM8 ectodomain Asp158-Ser653 Accession # P78325 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 8; a disintegrin and metalloproteinase domain 8 ADAM 8 ADAM metallopeptidase domain 8 CD156a CD156a antigen CD156human leukocyte differentiation antigen Cell surface antigen MS2 EC 3.4.24 EC 3.4.24.- MGC134985 MS2 MS2disintegrin and metalloproteinase domain-containing protein 8; ADAM 8; CD156a; Cell surface antigen MS2; Disintegrin and metalloproteinase domain-containing protein 8; human leukocyte differentiation antigen
Gene Aliases: ADAM8; CD156; CD156a; MS2
UniProt ID: (Human) P78325
Entrez Gene ID: (Human) 101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.