Proteintech
Manufacturer Code:237781AP150UL
Catalog # 50-173-4121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADAM8 Fusion Protein Ag17262 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 8; ADAM 8; ADAM metallopeptidase domain 8 ADAM8 ADAM8 protein CD156 MS2; CD156a; Cell surface antigen MS2; Disintegrin and metalloproteinase domain-containing protein 8; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein
Gene Aliases: ADAM 8; ADAM8; CD156; CD156a; E430039A18Rik; MS2
UniProt ID: (Human) P78325, (Mouse) Q05910
Entrez Gene ID: (Human) 101, (Mouse) 11501
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.