Invitrogen
Manufacturer Code:PA584536
Catalog # PIPA584536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ADAM32 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 32; a disintegrin and metalloproteinase domain 32; ADAM 32; Disintegrin and metalloproteinase domain-containing protein 32; metalloproteinase 12-like protein; testicular tissue protein Li 13
Gene Aliases: ADAM32; UNQ5982/PRO21340
UniProt ID: (Human) Q8TC27
Entrez Gene ID: (Human) 203102
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.