R&D Systems
Manufacturer Code:BAF5380
Catalog # BAF5380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ADAM32 Ser17-Thr476 Accession # Q8TC27 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 32; a disintegrin and metalloprotease domain 32 a disintegrin and metalloproteinase domain 32 ADAM 32 ADAM metallopeptidase domain 32 disintegrin and metalloproteinase domain-containing protein 32 FLJ26299 FLJ29004 metalloproteinase 12-like protein; a disintegrin and metalloproteinase domain 32; ADAM 32; Disintegrin and metalloproteinase domain-containing protein 32; metalloproteinase 12-like protein; testicular tissue protein Li 13
Gene Aliases: ADAM32; UNQ5982/PRO21340
UniProt ID: (Human) Q8TC27
Entrez Gene ID: (Human) 203102
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.