Invitrogen
Manufacturer Code:MA521251
Catalog # PIMA521251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E5 |
Immunogen | ADAM30 (NP&lowbar068566 199 a.a. &bsim 298 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 30; ADAM 30; ADAM metallopeptidase domain 30 svph4; Disintegrin and metalloproteinase domain-containing protein 30
Gene Aliases: ADAM30; svph4; UNQ2509/PRO5997
UniProt ID: (Human) Q9UKF2
Entrez Gene ID: (Human) 11085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.