Abnova
Manufacturer Code:PAB18828
Catalog # 89-120-387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of human ADAM30. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ADAM30.
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 30; ADAM 30; Disintegrin and metalloproteinase domain-containing protein 30; svph4
Gene Aliases: ADAM30; svph4; UNQ2509/PRO5997
UniProt ID: (Human) Q9UKF2
Entrez Gene ID: (Human) 11085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.