Invitrogen
Manufacturer Code:PA569435
Catalog # PIPA569435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ADAM29 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 29; a disintegrin and metalloproteinase domain 29; ADAM 29; Cancer/testis antigen 73; CT73; CT73 svph1; Disintegrin and metalloproteinase domain-containing protein 29; metallaproteinase-disintegrin (ADAM29); testis secretory sperm-binding protein Li 207a
Gene Aliases: ADAM29; CT73; svph1
UniProt ID: (Human) Q9UKF5
Entrez Gene ID: (Human) 11086, (Dog) 100687448, (Horse) 100065626, (Mouse) 244486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.