R&D Systems
Manufacturer Code:MAB9325100
Catalog # MAB9325100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2248A |
Immunogen | Chinese Hamster Ovary cell line CHO-derived human ADAM28 protein Leu72 & Leu191-Ala623 with a C-terminal 6-His tag Accession No. Q0UKQ2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
Protein Aliases: a disintegrin and metalloproteinase domain 28 ADAM metallopeptidase domain 28 ADAM23MDC-L disintegrin and metalloproteinase domain-containing protein 28 Dtgn1 EC 3.4.24 EC 3.4.24.- eMDC II eMDCII Epididymial metalloproteinase-like disintegrin-like and cysteine-rich proteinII MDCL MDCLADAM 28 MDC-Lm MDC-Ls Metalloproteinase-like disintegrin-like and cysteine-rich protein L metalloproteinase-like disintegrin-like and cysteine-rich protein-L Protein-L; ADAM 28; Disintegrin and metalloproteinase domain-containing protein 28; eMDC II; Epididymal metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; MDC-L; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L; metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L
Gene Aliases: ADAM 28; ADAM23; ADAM28; eMDC II; eMDCII; MDC-L; MDCL
UniProt ID: (Human) Q9UKQ2
Entrez Gene ID: (Human) 10863
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.