Novus Biologicals
Manufacturer Code:NBP267246
Catalog # NBP267246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Partial recombinant human ADAM28 protein (amino acids 72-623) [UniProt Q9UKQ2] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: a disintegrin and metalloproteinase domain 28 ADAM metallopeptidase domain 28 ADAM23MDC-L disintegrin and metalloproteinase domain-containing protein 28 EC 3.4.24 EC 3.4.24.- eMDC II eMDCII Epididymial metalloproteinase-like disintegrin-like and cysteine-rich proteinII MDCLADAM 28 MDC-Lm MDC-Ls Metalloproteinase-like disintegrin-like and cysteine-rich protein L metalloproteinase-like disintegrin-like and cysteine-rich protein-L; ADAM 28; Disintegrin and metalloproteinase domain-containing protein 28; eMDC II; Epididymal metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich protein II; MDC-L; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L; metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L
Gene Aliases: ADAM 28; ADAM23; ADAM28; eMDC II; eMDCII; MDC-L; MDCL
UniProt ID: (Human) Q9UKQ2
Entrez Gene ID: (Human) 10863
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.