Invitrogen
Manufacturer Code:PA568083
Catalog # PIPA568083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human ADAM23. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 23; ADAM 23; Disintegrin and metalloproteinase domain-containing protein 23; MDC-3; MDC3 MDC-3 disintegrin and metalloproteinase domain-containing protein 23 metalloproteinase-like disintegrin-like and cysteine-rich protein 3; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Gene Aliases: ADAM23; AW046396; MDC-3; MDC3
UniProt ID: (Human) O75077, (Mouse) Q9R1V7
Entrez Gene ID: (Human) 8745, (Mouse) 23792
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.