Novus Biologicals
Manufacturer Code:NBP215283
Catalog # NBP215283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ADAM23. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 23 ADAM metallopeptidase domain 23 disintegrin and metalloproteinase domain-containing protein 23 MDC-3 MDC3ADAM 23 Metalloproteinase-like disintegrin-like and cysteine-rich protein 3; ADAM 23; Disintegrin and metalloproteinase domain-containing protein 23; MDC-3; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Gene Aliases: ADAM23; MDC-3; MDC3
UniProt ID: (Human) O75077
Entrez Gene ID: (Human) 8745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.