Invitrogen
Manufacturer Code:MA524210
Catalog # PIMA524210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 498805 |
Immunogen | CHO-derived recombit human ADAM22 Gly26-His528 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 22; ADAM 22; ADAM 22 MDC2 a disintegrin and metalloproteinase domain 22 disintegrin and metalloproteinase domain-containing protein 22 metalloproteinase-disintegrin ADAM22-3 metalloproteinase-like disintegrin-like and cysteine-rich protein 2; Disintegrin and metalloproteinase domain-containing protein 22; Metalloproteinase-disintegrin ADAM22-3; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2
Gene Aliases: ADAM 22; ADAM22; MDC2
UniProt ID: (Human) Q9P0K1
Entrez Gene ID: (Human) 53616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.