Proteintech
Manufacturer Code:139521AP150UL
Catalog # 50-172-9116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADAM2 Fusion Protein Ag5063 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 2 (fertilin beta); ADAM 2; ADAM 2 ADAM metallopeptidase domain 2 ADAM2 Cancer/testis antigen 15 CRYN1 CRYN2 CT15 Fertilin subunit beta FTNB PH 30 PH 30b PH30 PH30 beta; Cancer/testis antigen 15; CT15; Disintegrin and metalloproteinase domain-containing protein 2; fertilin beta; Fertilin subunit beta; PH-30; PH30; PH30-beta
Gene Aliases: ADAM2; AI323749; CRYN1; CRYN2; CT15; FTNB; PH-30b; PH30; PH30-beta
UniProt ID: (Human) Q99965, (Mouse) Q60718
Entrez Gene ID: (Human) 2515, (Mouse) 11495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.