Novus Biologicals
Manufacturer Code:NBP289702100UL
Catalog # NB020885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 737 |
Immunogen | Recombinant Human ADAM12 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloproteinase domain 12 (meltrin alpha) ADAM 12 ADAM metallopeptidase domain 12 disintegrin and metalloproteinase domain-containing protein 12 EC 3.4.24 EC 3.4.24.58 MCMP MCMPMltna Meltrin-alpha MLTNA MLTNEC 3.4.24.-; ADAM 12; Disintegrin and metalloproteinase domain-containing protein 12; Meltrin-alpha; metalloprotease-disintegrin 12 transmembrane
Gene Aliases: ADAM12; ADAM12-OT1; CAR10; MCMP; MCMPMltna; MLTN; MLTNA; UNQ346/PRO545
UniProt ID: (Human) O43184
Entrez Gene ID: (Human) 8038
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.