R&D Systems
Manufacturer Code:IC1427F
Catalog # IC1427F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 163003 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ADAM10 Thr214-Glu672 Accession # O14672 |
Conjugate | Fluorescein |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a disintegrin and metalloprotease domain 10; a disintegrin and metalloprotease domain 10 a disintegrin and metalloproteinase domain 10 AD10 ADAM 10 ADAM metallopeptidase domain 10 CD156c CD156c antigen CDw156 disintegrin and metalloproteinase domain-containing protein 10 EC 3.4.24 HsT18717 kuz Kuzbanian Kuzbanian protein homolog MADM MADMEC 3.4.24.81 Mammalian disintegrin-metalloprotease; ADAM 10; CD156c; CDw156; Disintegrin and metalloproteinase domain-containing protein 10; Kuzbanian protein homolog; Mammalian disintegrin-metalloprotease
Gene Aliases: AD10; AD18; ADAM10; CD156c; CDw156; HsT18717; KUZ; MADM; RAK
UniProt ID: (Human) O14672
Entrez Gene ID: (Human) 102
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.