Invitrogen
Manufacturer Code:PA543731
Catalog # PIPA543731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ADAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine deaminase-like protein; Adenosine deaminase-like protein isoform 1; Adenosine deaminase-like protein-like protein; HsMAPDA; N6-mAMP deaminase; N6-methyl-AMP aminohydrolase
Gene Aliases: 4930578F03Rik; ADAL; ADAL1; zgc:109912
UniProt ID: (Human) Q6DHV7, (Bovine) Q0VC13, (Mouse) A2ARU3, (Zebrafish) Q4V9P6
Entrez Gene ID: (Dog) 100856728, (Human) 161823, (Pig) 100155388, (Horse) 100070813, (Bovine) 512667, (Rabbit) 100358454, (Guinea pig) 100724593, (Rat) 311352, (Mouse) 75894, (Zebrafish) 619263
Molecular Function:
deaminase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.