Proteintech
Manufacturer Code:212171AP150UL
Catalog # 50-173-3324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ADAD2 Fusion Protein Ag15650 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAD2 TENRL; adenosine deaminase domain containing 2; Adenosine deaminase domain-containing protein 2; Testis nuclear RNA-binding protein-like
Gene Aliases: 4930403J07Rik; ADAD2; RGD1560949; TENRL
UniProt ID: (Human) Q8NCV1, (Mouse) Q9D5P4
Entrez Gene ID: (Human) 161931, (Rat) 691275, (Mouse) 75773
Molecular Function:
DNA binding protein
RNA binding protein
deaminase
defense/immunity protein
enzyme modulator
hydrolase
kinase activator
kinase modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.