Novus Biologicals
Manufacturer Code:NBP299304FR
Catalog # NB083308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ACYP2 (P14621 Ser2-Tyr99). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acylphosphatase 2 muscle type acylphosphatase-2 Acylphosphate phosphohydrolase 2 ACYM ACYPmuscle type isozyme EC 3.6.1.7; acylphosphatase 2, muscle type; Acylphosphatase, muscle type isozyme; Acylphosphatase-2; Acylphosphate phosphohydrolase 2; testicular tissue protein Li 11
Gene Aliases: ACYM; ACYP; ACYP2
UniProt ID: (Human) P14621
Entrez Gene ID: (Human) 98
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.