Proteintech
Manufacturer Code:123021AP150UL
Catalog # 50-172-7875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACYP2 Fusion Protein Ag2948 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylphosphatase 2 acylphosphatase 2 muscle type ACYM ACYP ACYP2; acylphosphatase 2, muscle type; Acylphosphatase, muscle type isozyme; Acylphosphatase-2; Acylphosphate phosphohydrolase 2; testicular tissue protein Li 11
Gene Aliases: 2310004B09Rik; ACYM; ACYP; ACYP2
UniProt ID: (Human) P14621, (Mouse) P56375
Entrez Gene ID: (Human) 98, (Rat) 364224, (Mouse) 75572
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.