Novus Biologicals
Manufacturer Code:NBP290125C
Catalog # NB070133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human ACYP1 (P07311 Ala 2-Lys 99). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: acylphosphatase 1 erythrocyte (common) type Acylphosphatase erythrocyte isozyme Acylphosphatase organ-common type isozyme Acylphosphate phosphohydrolase 1 ACYPEacylphosphatase-1 EC 3.6.1.7; acylphosphatase 1, erythrocyte (common) type; Acylphosphatase, erythrocyte isozyme; Acylphosphatase, organ-common type isozyme; Acylphosphatase-1; Acylphosphate phosphohydrolase 1
Gene Aliases: ACYP1; ACYPE
UniProt ID: (Human) P07311
Entrez Gene ID: (Human) 97
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.