Invitrogen
Manufacturer Code:MA530037
Catalog # PIMA530037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 37 |
Immunogen | Recombit Human ACVR2B Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IIB; activin A receptor, type IIB; Activin receptor type IIB; Activin receptor type-2B; ACTR-IIB; ACTRIIB ActR-IIB MGC116908
Gene Aliases: ActR-IIB; ACTRIIB; ACVR2B; HTX4
UniProt ID: (Human) Q13705
Entrez Gene ID: (Human) 93
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.