Invitrogen
Manufacturer Code:PA5111122
Catalog # PIPA5111122
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ACVR2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IIB; activin A receptor, type IIB; Activin receptor type IIB; Activin receptor type IIB Activin receptor type-2B Actriib ACTR-IIB Acvr2b HTX4 MGC116908; Activin receptor type-2B; ACTR-IIB
Gene Aliases: ActR-IIB; ACTRIIB; ACVR2B; HTX4
UniProt ID: (Human) Q13705
Entrez Gene ID: (Human) 93
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.