Invitrogen
Manufacturer Code:PA595374
Catalog # PIPA595374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human ACVR2A recombit protein (Position: Q421-L513). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IIA; activin A receptor, type IIA; activin receptor IIA; Activin receptor type IIA; Activin receptor type-2A; Activin receptor type-2A 2.7.11.30 Activin receptor type IIA ACTR-IIA ACTRIIA ACVR2A ACVR2; ACTR-IIA; type II activin receptor
Gene Aliases: ACTRII; ACVR2; ACVR2A; rActR-II
UniProt ID: (Human) P27037, (Rat) P38444
Entrez Gene ID: (Human) 92, (Rat) 29263
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.