Invitrogen
Manufacturer Code:PA513886
Catalog # PIPA513886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 2-29 amino acids from the N-terminal region of human ACVR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IIA; activin A receptor, type IIA; Activin receptor type IIA; Activin receptor type-2A; Activin receptor type-2A &lsqbPrecursor] Activin receptor type IIA ACTR-IIA ACTRIIA ACVR2; ACTR-IIA
Gene Aliases: ACTRII; ActrIIa; ACVR2; ACVR2A; TactrII
UniProt ID: (Human) P27037, (Mouse) P27038
Entrez Gene ID: (Human) 92, (Mouse) 11480
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.