Proteintech
Manufacturer Code:126101AP150UL
Catalog # 50-172-8111
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACVR1C Fusion Protein Ag3162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type IC; activin A receptor type IC Activin receptor like kinase 7 Activin receptor type 1C Activin receptor type IC ACTR IC ACVR1C ACVRLK7 ALK 7 ALK7; activin A receptor, type IC; Activin receptor type IC; Activin receptor type-1C; Activin receptor-like kinase 7; ACTR-IC; ACVR1C; acvr1c {ECO:0000312|EMBL:AAC52803.1}; ALK-7; TGF-beta type 1 receptor
Gene Aliases: ACVR1C; ACVRLK7; Alk-7; ALK7; C230097P10; habrec1
UniProt ID: (Human) Q8NER5, (Rat) P70539, (Mouse) Q8K348
Entrez Gene ID: (Human) 130399, (Rat) 245921, (Mouse) 269275
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.