Invitrogen
Manufacturer Code:MA538039
Catalog # PIMA538039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1899 |
Immunogen | A synthesized peptide derived from human ACVR1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 6820432J04 activin A receptor type 1B activin A receptor type IB activin A receptor type 1B activin A receptor type IB activin A receptor type II-like kinase 4 activin receptor type IB Activin receptor type-1B activin receptor-like kinase 4 ACTRI ActRIB ActR-IB ACVR1A ACVR1B ACVRLK2 Acvrlk4 ALK2 Alk4 ALK-4 FOP Serine/threonine-protein kinase receptor R2 SKR1 SKR2 TSRI; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; Activin receptor type IB; Activin receptor type-1B; Activin receptor-like kinase 4; ACTR-IB; ALK-4; Serine/threonine-protein kinase receptor R2; SKR2
Gene Aliases: 6820432J04; ActR-IB; ACTRIB; ACVR1B; ACVRLK4; ALK4; SKR2
UniProt ID: (Human) P36896, (Rat) P80202, (Mouse) Q61271
Entrez Gene ID: (Human) 91, (Rat) 29381, (Mouse) 11479
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.