Abnova
Manufacturer Code:PAB3465
Catalog # 89-131-028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ACVR1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ACVR1B.
For Research Use Only
Protein Aliases: activin A receptor type IB; activin A receptor, type IB; activin A receptor, type II-like kinase 4; Activin receptor type IB; Activin receptor type-1B; Activin receptor-like kinase 4; ACTR-IB; ACTRIB; ACVRLK4; ALK-4; ALK4; Serine/threonine-protein kinase receptor R2; SKR2
Gene Aliases: ACTRIB; ACVR1B; ACVRLK4; ALK4; SKR2
UniProt ID: (Human) P36896
Entrez Gene ID: (Human) 91
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.