Invitrogen
Manufacturer Code:MA537901
Catalog # PIMA537901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2449 |
Immunogen | Recombinant protein of human ACVR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type 1 activin A receptor type I activin A receptor type 1 activin A receptor type I activin A receptor type II-like kinase 2 activin receptor type I Activin receptor type-1 activin receptor-like kinase 2 activin type I receptor ACTRI ACTR-I ActRIA Acvr Acvr1 ACVR1A ACVRLK2 ALK2 ALK-2 ALK2-B Alk8 D330013D15Rik FOP hydroxyalkyl-protein kinase serine/threonine-protein kinase receptor R1 SKR1 TGF-B superfamily receptor type I Tgfb1 Tsk7L TSK-7L TSRI TSR-I Type I TGF B receptor; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; Activin receptor type I; Activin receptor type-1; Activin receptor-like kinase 2; activin type I receptor; ACTR-I; ALK-2; ALK2-B; hydroxyalkyl-protein kinase; Serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; TSK-7L; TSR-I
Gene Aliases: ActR-I; ACTRI; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; Alk-2; ALK2; Alk8; D330013D15Rik; FOP; SKR1; Tgfb1; Tsk7L; TSRI
UniProt ID: (Human) Q04771, (Rat) P80201, (Mouse) P37172
Entrez Gene ID: (Human) 90, (Rat) 79558, (Mouse) 11477
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.