Invitrogen
Manufacturer Code:PA528662
Catalog # PIPA528662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 214 and 509 of Human Activin A receptor type I |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; Activin receptor type I; Activin receptor type-1; Activin receptor-like kinase 2; activin type I receptor; ACTR-I; ACTRI ACVR1A ACVRLK2 ALK2 FOP SKR1 TSRI TGF-B superfamily receptor type I activin A receptor type II-like kinase 2 activin receptor type I activin receptor type-1 activin receptor-like kinase 2 hydroxyalkyl-protein kinase serine/threonine-protein kinase receptor R1; ALK-2; ALK2-B; hydroxyalkyl-protein kinase; Serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; TSR-I
Gene Aliases: ACTRI; ACVR1; ACVR1A; ACVRLK2; ALK2; FOP; SKR1; TSRI
UniProt ID: (Human) Q04771, (Rat) P80201
Entrez Gene ID: (Human) 90, (Rat) 79558
Molecular Function: TGF-beta receptor cytokine receptor kinase protein kinase protein kinase receptor receptor serine/threonine protein kinase receptor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.