Invitrogen
Manufacturer Code:PA518842
Catalog # PIPA518842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RKFKRRNQERLNPRD) corresponding to the internal amino acids of ACVR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activin A receptor type I; activin A receptor, type I; activin A receptor, type II-like kinase 2; Activin receptor type I; Activin receptor type-1; Activin receptor-like kinase 2; ACTR-I; ACVR1 activin A receptor type I ACTRI ACVRLK2 ALK2 FOP SKR1 activin A receptor type II-like kinase 2 activin A type I receptor hydroxyalkyl-protein kinase; ALK-2; hydroxyalkyl-protein kinase; Serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; TSR-I
Gene Aliases: ACTRI; ACVR1; ACVR1A; ACVRLK2; ALK2; FOP; SKR1; TSRI
UniProt ID: (Human) Q04771
Entrez Gene ID: (Human) 90
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.