Abnova
Manufacturer Code:PAB9840
Catalog # 89-134-445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to extracellular region of chicken ACVR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against partial recombinant ACVR1.
For Research Use Only
Protein Aliases: activin A receptor type 1; activin A receptor, type I; Activin receptor type I; Activin receptor type-1; Type I TGF B receptor
Gene Aliases: ACVR1; ALK2
UniProt ID: (Chicken) Q90ZK6
Entrez Gene ID: (Chicken) 395246
Molecular Function:
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.