Invitrogen
Manufacturer Code:PA5109359
Catalog # PIPA5109359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 237-396 of human ACTR6 (NP_071941.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010200J04Rik Actin-related protein 6 Actr6 ARP6 ArpX AU044443 BOS_5280 CDA12 FLJ13433 h hARPX hArp6 hARPX HSPC281 mArp6 MSTP136; Actin-related protein 6; ARP6 actin-related protein 6 homolog; hArp6; hARPX; mArp6
Gene Aliases: 2010200J04Rik; ACTR6; ARP6; ArpX; AU044443; CDA12; hARP6; hARPX; HSPC281; MSTP136
UniProt ID: (Human) Q9GZN1, (Mouse) Q9D864
Entrez Gene ID: (Human) 64431, (Rat) 314718, (Mouse) 67019
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.