Novus Biologicals
Manufacturer Code:NBP2843920.1ML
Catalog # NB032320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human ACTR6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-related protein 6; actin-related protein 6 ARP6 ARP6 actin-related protein 6 homolog (yeast) FLJ13433 hARP6 hARPX HSPC281 MSTP136; hArp6; hARPX
Gene Aliases: ACTR6; ARP6; CDA12; hARP6; hARPX; HSPC281; MSTP136
UniProt ID: (Human) Q9GZN1
Entrez Gene ID: (Human) 64431
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.