Invitrogen
Manufacturer Code:PA526274
Catalog # PIPA526274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 539-568 amino acids from the C-terminal region of human ACTR5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-related protein 5; ARP5 actin-related protein 5 homolog (yeast); Arp5 INO80M INO80 complex subunit M actin-related protein 5 hARP5 sarcoma antigen NY-SAR-16; hARP5; INO80 complex subunit M; Sarcoma antigen NY-SAR-16
Gene Aliases: ACTR5; ARP5; I79_002649; INO80M
UniProt ID: (Human) Q9H9F9
Entrez Gene ID: (Human) 79913, (Chinese hamster) 100765489
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.