Novus Biologicals
Manufacturer Code:NBP303979
Catalog # NB093221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ACTR5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-related protein 5; ACTR5 ARP5 actin-related protein 5 homolog (yeast) Arp5 INO80M; hARP5; INO80 complex subunit M; Sarcoma antigen NY-SAR-16
Gene Aliases: ACTR5; ARP5; INO80M
UniProt ID: (Human) Q9H9F9
Entrez Gene ID: (Human) 79913
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.