Invitrogen
Manufacturer Code:PA544774
Catalog # PIPA544774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ACTR3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-like protein 3B; actin-related protein 3-beta; Actin-related protein 3B; actin-related protein Arp11; Actin-related protein ARP4; ARP3 actin related protein 3 homolog B; ARP3 actin-related protein 3 homolog B; ARP3 actin-related protein 3 homolog B (yeast) ARP11 ARP3BETA DKFZp686O24114; ARP3-beta
Gene Aliases: 9630005C02; ACTR3B; ARP11; Arp3b; ARP3BETA; ARP4; AW047569; RGD1565759; zgc:158823
UniProt ID: (Human) Q9P1U1, (Mouse) Q641P0
Entrez Gene ID: (Horse) 100065073, (Rabbit) 100343474, (Dog) 475549, (Human) 57180, (Mouse) 242894, (Rat) 362298, (Zebrafish) 100038776
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.