Novus Biologicals
Manufacturer Code:NBP261806
Catalog # NBP2-61806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G2C9 |
Immunogen | Purified recombinant fragment of human ACTR3 (AA: 287-418) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: Actin-like protein 3; Actin-related protein 3; actin-related protein 3 ARP3 (actin-related protein 3 yeast) homolog ARP3 actin-related protein 3 homolog (yeast) ARP3Actin-like protein 3; ARP3 actin-related protein 3 homolog
Gene Aliases: ACTR3; ARP3
UniProt ID: (Human) P61158
Entrez Gene ID: (Human) 10096
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.