Invitrogen
Manufacturer Code:PA530355
Catalog # PIPA530355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 181 of Human Beta centractin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-related protein 1B; ARP1 actin-related protein 1 homolog B, centractin beta; ARP1B; ARP1B CTRN2 PC3 actin-related protein 1B beta-centractin centractin beta; Beta-centractin; centractin beta
Gene Aliases: 2310066K23Rik; AA960180; ACTR1B; AI851923; ARP1B; CTRN2; PC3
UniProt ID: (Human) P42025, (Mouse) Q8R5C5
Entrez Gene ID: (Human) 10120, (Mouse) 226977
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.