Novus Biologicals
Manufacturer Code:NBP215275
Catalog # NBP215275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ACTR1B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-related protein 1B; Actin-related protein 1B ARP1 (actin-related protein 1 yeast) homolog B (centractin beta) ARP1 actin-related protein 1 homolog B centractin beta (yeast) ARP1BPC3 centractin beta CTRN2beta-centractin; ARP1B; Beta-centractin; centractin beta
Gene Aliases: ACTR1B; ARP1B; CTRN2; PC3
UniProt ID: (Human) P42025
Entrez Gene ID: (Human) 10120
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.